Learn More
Alkali Scientific 96 Multi-Well Culture Plate, Non-Treated for Suspension Culture, PS, Black, Flat bottom, 50/cs
SDP
Catalog No. 502389199
Encompass_Preferred
ASI™ 96 Well Culture Solid Black Plates have a low auto-fluorescence with efficient light blocking ability to obtain most accurate result in fluorescence experiments. Black microplates have low background fluorescence and minimize light scattering. ▪ High Throughput Screening compatible ▪ Suitable for fluorescence & luminescence assays ▪ Non treated for suspension cultures ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Molded Alphanumeric labeling ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant ▪ Non pyrogenic ▪ Non cytotoxic ▪ DNase / RNase / Pyrogen free ▪ Human DNA free
Catalog No. 50-238-9199 Supplier Alkali Scientific Supplier No. TPB96
ASI™ 96 Well Culture Solid Black Plates have a low auto-fluorescence with efficient light blocking ability to obtain most accurate result in fluorescence experiments. Black microplates have low background fluorescence and minimize light scattering. ▪ High Throughput Screening compatible ▪ Suitable for fluorescence & luminescence assays ▪ Non treated for suspension cultures ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Molded Alphanumeric labeling ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant ▪ Non pyrogenic ▪ Non cytotoxic ▪ DNase / RNase / Pyrogen free ▪ Human DNA free