Learn More
Electron Microscopy Sciences Oncyte Microtiter Plates Nova, 96 square, 6x6 mm I.D.
SDP
Catalog No. 5027553
Encompass_Preferred
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. NOVA for highest fluorescent sensitivity, reduced binding. SBS Compliant w/Glass Substrate.
Wells: 96
I.D.: 6 x 6 mm
Catalog No. 50-275-53 Supplier Electron Microscopy Sciences Supplier No. 7033571N
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. NOVA for highest fluorescent sensitivity, reduced binding. SBS Compliant w/Glass Substrate.
Wells: 96
I.D.: 6 x 6 mm