Learn More
Waters Corp Connected Device, 96-PCR Magnet+, Automated Moving Shelf, Magnetic Bead-Based Purification
Catalog No. 502390646 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 96-PCR Magnet+ is a connected device designed to simplify and ensure reliability in the purification of genomic, plasmid, and mitochondrial DNA using magnetic beads. It features an automated moving shelf mechanism that strategically positions magnets near the bottom sides of consumables, optimizing the process of bead pelleting.
  • Ensures a simple and reliable method of purifying DNA.
  • Utilizes magnetic beads for purification.
  • Automated moving shelf strategically positions magnets.
  • Optimizes bead pelleting in consumables.
Catalog No. 50-239-0646 Supplier Waters Corp Supplier No. 176004850
Add to Cart
Add to Cart
Request a Quote
The 96-PCR Magnet+ is a connected device designed to simplify and ensure reliability in the purification of genomic, plasmid, and mitochondrial DNA using magnetic beads. It features an automated moving shelf mechanism that strategically positions magnets near the bottom sides of consumables, optimizing the process of bead pelleting.
  • Ensures a simple and reliable method of purifying DNA.
  • Utilizes magnetic beads for purification.
  • Automated moving shelf strategically positions magnets.
  • Optimizes bead pelleting in consumables.