Learn More
Andwin Scientific 96-PLACE RACK FOR 0.2 ML TUBES
SDP
Catalog No. 50819912
Encompass_Preferred
The polypropylene 96-place rack can store 0.2 ml individual and strip tubes. It can endure autoclaving and freezing temperatures of down to -90C. The transparent lid allows the racks to be stacked, which then permits the user to efficiently save space. It is sold in a pack of 5 racks in assorted neon colors of blue, green, orange, pink, and yellow with 5 natural lids.
Catalog No. 50-819-912 Supplier Andwin Scientific Supplier No. 23009602
The polypropylene 96-place rack can store 0.2 ml individual and strip tubes. It can endure autoclaving and freezing temperatures of down to -90C. The transparent lid allows the racks to be stacked, which then permits the user to efficiently save space. It is sold in a pack of 5 racks in assorted neon colors of blue, green, orange, pink, and yellow with 5 natural lids.