Learn More
Waters Corp Waters 96 Place Sealing Gasket, for Positive Pressure 96 Processor
Catalog No. 50466132 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-place sealing gasket is designed for positive pressure processors to maintain uniform pressure across all 96 holes in the manifold. It is a critical component for ensuring consistent solid-phase extraction by providing restricted output of inert gas, even when not all plate well positions are filled.
  • Maintains constant pressure across all 96 holes in the processor’s manifold
  • Ensures uniform solid-phase extraction (SPE)
  • Provides restricted output of inert gas at every port
  • Effective even if not all plate well positions are filled
  • Has a shelf life of 6 months from the shipping date
  • Requires special storage at 4°C
  • Can be cleaned using a soft cloth dampened with methanol
Catalog No. 50-466-132 Supplier Waters Corp Supplier No. 186005522
This 96-place sealing gasket is designed for positive pressure processors to maintain uniform pressure across all 96 holes in the manifold. It is a critical component for ensuring consistent solid-phase extraction by providing restricted output of inert gas, even when not all plate well positions are filled.
  • Maintains constant pressure across all 96 holes in the processor’s manifold
  • Ensures uniform solid-phase extraction (SPE)
  • Provides restricted output of inert gas at every port
  • Effective even if not all plate well positions are filled
  • Has a shelf life of 6 months from the shipping date
  • Requires special storage at 4°C
  • Can be cleaned using a soft cloth dampened with methanol