Learn More
Arcticwhite LLC 96 Round Deep BioBlock Well, 1 ML, Polypropylene, Round Bottom, Natural, 24 Microplates per Case
SDP
Catalog No. NC9159750
Encompass
96 Round Deep BioBlock Well, 1 ML/Well, Polypropylene, Round Bottom, Natural, 24 Microplates per Case. The nominal well volume is 1 mL. Each well holds up to 1.2 mL.Use for sample archiving and automated workflows. Supplied by ArcticWhite as 24 microplates per case.
Catalog No. NC9159750 Supplier Arcticwhite LLC Supplier No. AWLST1105
96 Round Deep BioBlock Well, 1 ML/Well, Polypropylene, Round Bottom, Natural, 24 Microplates per Case. The nominal well volume is 1 mL. Each well holds up to 1.2 mL.Use for sample archiving and automated workflows. Supplied by ArcticWhite as 24 microplates per case.