Learn More
Arcticwhite LLC 96 Square Deep Well, 2 ML, Polypropylene, Pyramid Bottom, Standard / 25/Case, 96-well, ANSI/SLAS (SBS) Standard Footprint, 2 ML per Well, Medical-grade Polypropylene (PP), Chemical Resistant, 25 Deep Well Plates per Case
SDP
Catalog No. NC0397239
Encompass
96 Square Deep Well, 2 ML, Polypropylene, Pyramid Bottom, Standard / 25/Case, 96-well, ANSI/SLAS (SBS) Standard Footprint, 2 ML per Well, Medical-grade Polypropylene (PP), Chemical Resistant This 96-well deep well microplate is used for automated liquid handling, nucleic acid purification, cell and bacterial culture, and sample storage. Supplied by ArcticWhite as 25 deep well plates per case.
Catalog No. NC0397239 Supplier Arcticwhite LLC Supplier No. AWLSS30009
96 Square Deep Well, 2 ML, Polypropylene, Pyramid Bottom, Standard / 25/Case, 96-well, ANSI/SLAS (SBS) Standard Footprint, 2 ML per Well, Medical-grade Polypropylene (PP), Chemical Resistant This 96-well deep well microplate is used for automated liquid handling, nucleic acid purification, cell and bacterial culture, and sample storage. Supplied by ArcticWhite as 25 deep well plates per case.