Learn More
Arcticwhite LLC 96 Square Deep Well, 350 UL/Well, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS Base and Footprint, Square Well, Conical Bottom, Polypropylene
SDP
Catalog No. NC3735320
Encompass
96 Square Deep Well, 350 UL/Well, Polypropylene, Conical Bottom. A deep well microplate for sample storage and processing. 96 Well Format, ANSI/SLAS standard base and footprint. Square well shape. Conical Bottom. 350 uL Well volume. Polypropylene Material.
Catalog No. NC3735320 Supplier Arcticwhite LLC Supplier No. AWLS219006
96 Square Deep Well, 350 UL/Well, Polypropylene, Conical Bottom. A deep well microplate for sample storage and processing. 96 Well Format, ANSI/SLAS standard base and footprint. Square well shape. Conical Bottom. 350 uL Well volume. Polypropylene Material.