Learn More
Electron Microscopy Sciences ProPlate™ 96 Square FlexPlate, Adhesive 1 Side, 7.25x7.25 mm
SDP
Catalog No. 5034343
Encompass_Preferred
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. 7.25x7.25mm, Adhesive One Slide - Bottomless. In a removable well format.
Catalog No. 50-343-43 Supplier Electron Microscopy Sciences Supplier No. 6348510
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. 7.25x7.25mm, Adhesive One Slide - Bottomless. In a removable well format.