Learn More
Electron Microscopy Sciences ProPlate™ 96 Square FlexPlate, Adhesive 1 Side, 7.25x7.25mm Black
SDP
Catalog No. 5034344
Encompass_Preferred
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. 7.25x7.25mm, Black Polystyrene Frame, Adhesive One Slide - Bottomless In a removable well format.
Catalog No. 50-343-44 Supplier Electron Microscopy Sciences Supplier No. 6348511
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. 7.25x7.25mm, Black Polystyrene Frame, Adhesive One Slide - Bottomless In a removable well format.