Learn More
Thomson Instrument Company 96-Well Plate, 2mL, Square Well, Round Bottom, Rim, Skirted, Raised Letters
SDP
Catalog No. NC9241159
Encompass_Preferred
Thomson Well Plates come in 6-, 24-, and 96-well formats with multiple well shapes and bottoms for screening, sample prep, or concentration. They’re ANSI/SLAS compatible and offered with matching seals and lids for clean, repeatable workflows and automation.
•Format & geometry options — square or round wells; pyramid, round, or V-bottoms to match your assay and culture needs.
•Automation-friendly fixed plate orientation for liquid handlers.
•Sealing ecosystem — capmats, gas-permeable airporous seals, foil seals, and lids.
•Collection-plate role — used as the receiving plate beneath filter plates in clarification workflows.
• Made in the USA
This item is not returnable. View return policy
Catalog No. NC9241159 Supplier Thomson Instrument Company Supplier No. 951652B
Add to Cart
Add to Cart
Thomson Well Plates come in 6-, 24-, and 96-well formats with multiple well shapes and bottoms for screening, sample prep, or concentration. They’re ANSI/SLAS compatible and offered with matching seals and lids for clean, repeatable workflows and automation.
•Format & geometry options — square or round wells; pyramid, round, or V-bottoms to match your assay and culture needs.
•Automation-friendly fixed plate orientation for liquid handlers.
•Sealing ecosystem — capmats, gas-permeable airporous seals, foil seals, and lids.
•Collection-plate role — used as the receiving plate beneath filter plates in clarification workflows.
• Made in the USA