Learn More
Alkali Scientific Tip Comb, 96, V-Bottom, PP, for KingFisher™ and IsoPURE™, 50 bags of 2 Pieces, 100/cs
SDP
Catalog No. 502335769
Encompass_Preferred
Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc.
Catalog No. 50-233-5769 Supplier Alkali Scientific Supplier No. TL457
Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc.