Learn More
Quality Biological Inc 96 Well, Flat Bottom, Non-treated Plate without Lid, Individual, Sterile, 100/CS
SDP
Catalog No. 50147465
Encompass_Preferred
96 Well, Flat Bottom, Polystyrene, Non-treated Plate without Lid, Individual, Sterile
Packaged Individually, 50/PK, 100/CS
• Plates are labeled with alpha-numeric markings for easy identification of individual wells
• Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination
• Lids have a stacking feature for added stability when handling tall stacks of plates
• Working volume: 50-300 μL
Catalog No. 50-147-465 Supplier Quality Biological Inc Supplier No. P886229592
Add to Cart
Add to Cart
96 Well, Flat Bottom, Polystyrene, Non-treated Plate without Lid, Individual, Sterile
Packaged Individually, 50/PK, 100/CS
• Plates are labeled with alpha-numeric markings for easy identification of individual wells
• Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination
• Lids have a stacking feature for added stability when handling tall stacks of plates
• Working volume: 50-300 μL