Learn More
Quality Biological Inc 96 Well, Round Bottom, Non-treated Platewithout Lid, Individual, Sterile, 100/CS
SDP
Catalog No. 50147464
Encompass_Preferred
96 Well, Round Bottom, Polysytrene, Non-treated Platewithout Lid, Sterile
Packaged Individually, 50/PK, 100/CS
• Plates are labeled with alpha-numeric markings for easy identification of individual wells
• Round bottom wells for efficient mixing
• Packaged without lid
Catalog No. 50-147-464 Supplier Quality Biological Inc Supplier No. P886229591
96 Well, Round Bottom, Polysytrene, Non-treated Platewithout Lid, Sterile
Packaged Individually, 50/PK, 100/CS
• Plates are labeled with alpha-numeric markings for easy identification of individual wells
• Round bottom wells for efficient mixing
• Packaged without lid