Learn More
USA Scientific Inc TempPlate semi-skirted 96-well PCR plate, raised sides, natural
Catalog No. 507541657
Encompass_Preferred
TempPlate semi-skirted polypropylene 0.2 mL 96-well PCR plate, contoured and raised sides, standard depth, natural. Two/sleeve, 10/box.
Catalog No. 50-754-1657 Supplier USA Scientific Inc Supplier No. 14029300
TempPlate semi-skirted polypropylene 0.2 mL 96-well PCR plate, contoured and raised sides, standard depth, natural. Two/sleeve, 10/box.