Learn More
Thomas Scientific 96 Well Polystyrene, Clear, Flat Bottom, 350uL
SDP
Catalog No. NC1610787
Encompass_Preferred
For routine adsorption, absorption, ELISA, mixing and storage applications. The standard range of 96-well assay plates offers the perfect mix of affordability and high quality. Manufactured from high quanlity crystal polystyrene Flat bottom for spectrophotometric work V-bottom minimizing residual liquid Round (U) bottom for cell/particulate collection Robot compatible Working volumes from 275uL down to 10uL
Catalog No. NC1610787 Supplier Thomas Scientific Supplier No. 1167V76
For routine adsorption, absorption, ELISA, mixing and storage applications. The standard range of 96-well assay plates offers the perfect mix of affordability and high quality. Manufactured from high quanlity crystal polystyrene Flat bottom for spectrophotometric work V-bottom minimizing residual liquid Round (U) bottom for cell/particulate collection Robot compatible Working volumes from 275uL down to 10uL