Learn More
Research Products International Corp 96 Well 2-Tier Insert for Vortex-Genie 2
SDP
Catalog No. 501978109
Encompass
High-density, double layered foam insert for prolonged shaking of one standard footprint microplate to protect sample from heat-buildup. This insert fits into the 6 inch (152mm) Platform attachment for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Catalog No. 50-197-8109 Supplier Research Products International Corp Supplier No. 155303
High-density, double layered foam insert for prolonged shaking of one standard footprint microplate to protect sample from heat-buildup. This insert fits into the 6 inch (152mm) Platform attachment for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.