Learn More
Arcticwhite LLC 96 Round Deep Well, 1.6 ML, Polypropylene, Round Bottom, 96-well Plate, SBS Standard Footprint, 1.7 ML Maximum per Well, Medical-grade Polypropylene (PP), Storage Down to -86 Deg C, SBS-compliant, 50 Deep Well Plates per Case
SDP
Catalog No. NC9436665
Encompass
96 Round Deep Well, 1.6 ML/Well, Polypropylene, Round Bottom, 96-well Plate, SBS Standard Footprint, 1.7 ML Maximum per Well, Medical-grade Polypropylene (PP), Storage Down to -86 Deg C, 50 Deep Well Plates per Case. This 96-well deep well microplate is made from polypropylene and is used for automated liquid handling, nucleic acid purification, and cell or bacterial culture. Working volume - 1.6 mL per well. Volume when capped - 1.5 mL per well. Material - medical-grade polypropylene (PP). Chemical resistance - resistant to common laboratory chemicals. Thermal range - storage down to -86 deg C. Purity - DNase-free, RNase-free. Certifications - SBS-compliant. Supplied by ArcticWhite as 50 deep well plates per case.
Catalog No. NC9436665 Supplier Arcticwhite LLC Supplier No. AWLSX10005
96 Round Deep Well, 1.6 ML/Well, Polypropylene, Round Bottom, 96-well Plate, SBS Standard Footprint, 1.7 ML Maximum per Well, Medical-grade Polypropylene (PP), Storage Down to -86 Deg C, 50 Deep Well Plates per Case. This 96-well deep well microplate is made from polypropylene and is used for automated liquid handling, nucleic acid purification, and cell or bacterial culture. Working volume - 1.6 mL per well. Volume when capped - 1.5 mL per well. Material - medical-grade polypropylene (PP). Chemical resistance - resistant to common laboratory chemicals. Thermal range - storage down to -86 deg C. Purity - DNase-free, RNase-free. Certifications - SBS-compliant. Supplied by ArcticWhite as 50 deep well plates per case.