Learn More
Light Labs 96-well Aluminum Cooler Blocks Red
Catalog No. NC1624025
Encompass_Preferred
Keep PCR Tubes, Strip Tubes, PCR Plates, microtubes and cryovials cold on the bench top. Non-Skid rubber feet provide a stable platform for loading samples. Gets cold in as few as 25 minutes. Samples stay cold on your bench top for up to an hour. Small footprint minimizes bench top and freezer space used. Precisely drilled wells allow for maximum heat transfer between the block and samples.
Catalog No. NC1624025 Supplier Light Labs Supplier No. A7079R
Keep PCR Tubes, Strip Tubes, PCR Plates, microtubes and cryovials cold on the bench top. Non-Skid rubber feet provide a stable platform for loading samples. Gets cold in as few as 25 minutes. Samples stay cold on your bench top for up to an hour. Small footprint minimizes bench top and freezer space used. Precisely drilled wells allow for maximum heat transfer between the block and samples.