Learn More
Light Labs Aluminum Cooler Blocks, 96 Well, Silver
Catalog No. NC9820161
Encompass_Preferred
96-Well Aluminum Cooler Blocks, 1/ea Keep PCR Tubes, Strip Tubes, PCR Plates, microtubes and cryovials cold on the bench top. Precisely drilled wells allow for maximum heat transfer between the block and samples.
Catalog No. NC9820161 Supplier Light Labs Supplier No. A7079
96-Well Aluminum Cooler Blocks, 1/ea Keep PCR Tubes, Strip Tubes, PCR Plates, microtubes and cryovials cold on the bench top. Precisely drilled wells allow for maximum heat transfer between the block and samples.