Learn More
Research Products International Corp RPI 96-Well Assay Plates, Sterile, Round Bottom with Lid, Individually Wrapped, 100 per Case
SDP
Catalog No. 501367500
Encompass
Plates for high throughput screening and suspension culture applications. Plates are compatible with all popular high throughput sequencers. Suggested 40 - 280 μl working well volume. Plate ridges permit stable stacking to save bench top and incubator. Alphanumeric notation system for easy orientation and sample identification. Supplied as clear plastic. 
Catalog No. 50-136-7500 Supplier Research Products International Corp Supplier No. 141382
Add to Cart
Add to Cart
Plates for high throughput screening and suspension culture applications. Plates are compatible with all popular high throughput sequencers. Suggested 40 - 280 μl working well volume. Plate ridges permit stable stacking to save bench top and incubator. Alphanumeric notation system for easy orientation and sample identification. Supplied as clear plastic. 