Learn More
Analytical Sales & Services 96 Well, Black/Clear Flat Bottom PS Microplate, 25-340µL working volume per well
Catalog No. 5000804288
Encompass
96-Well Microplate\n\n• Polystyrene\n• Black with Clear Bottom\n• Flat Bottom\n• 25-340µL working volume per well\n• High Binding\n• Sterile\n\nQty/Pkg: 40
Catalog No. 50-008-04288 Supplier Analytical Sales & Services Supplier No. 96097
96-Well Microplate\n\n• Polystyrene\n• Black with Clear Bottom\n• Flat Bottom\n• 25-340µL working volume per well\n• High Binding\n• Sterile\n\nQty/Pkg: 40