Learn More
Zymo Research Corporation 96-Well Block, 10 Blocks
SDP
Catalog No. 501251650
Encompass_Preferred
96-Well Block w/ Cover Foil feature durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Catalog No. 50-125-1650 Supplier Zymo Research Corporation Supplier No. P100110
96-Well Block w/ Cover Foil feature durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.