Learn More
Zymo Research Corporation 96-Well Block w/ Cover Foil, 2 Blocks/foils
SDP
Catalog No. 502037211
Encompass_Preferred
96-Well Block w/ Cover Foil feature durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Catalog No. 50-203-7211 Supplier Zymo Research Corporation Supplier No. P10022
96-Well Block w/ Cover Foil feature durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.