Learn More
Thomas Scientific 96 Well Flexi PCR Plate
SDP
Catalog No. NC0020124
Encompass
Polypropylene 96 well Flexi plate that can be separated without scissors into 8,16,24,32,40,48,56,64,72,80,88,96 well Format, Certified RNase, DNase and Human DNA Free,Compatible with all major thermal cyclers.
Catalog No. NC0020124 Supplier Thomas Scientific Supplier No. 1207H41
Polypropylene 96 well Flexi plate that can be separated without scissors into 8,16,24,32,40,48,56,64,72,80,88,96 well Format, Certified RNase, DNase and Human DNA Free,Compatible with all major thermal cyclers.