Learn More
Thomson Instrument Company 96-Well Capmat, For Square Wells | Use w/ 931130
SDP
Supplier:  Thomson Instrument Company A210100
Encompass_Preferred
Thomson Instrument Company 96-Well Capmat for round wells is ideal for long-term storage in well plates. Thomson well plates are offered in 24 and 96-well configurations with a V, U or pyramid shape bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Many plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. NC3059855