Learn More
Chemglass Life Sciences 96-Well Cell Culture Plate, Treated, Flat, Sterile
SDP
Catalog No. 501897622
Encompass
• Made from high clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum Plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Non-pyrogenic, DNase- and RNase-free
Catalog No. 50-189-7622 Supplier Chemglass Life Sciences Supplier No. CGE3503096
• Made from high clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum Plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Non-pyrogenic, DNase- and RNase-free