Learn More
NEST SCIENTIFIC USA 96 Well Cell Culture Plate, flat, TC, sterile 100/cs
Catalog No. NC3429285
Encompass_Preferred
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. This part number comes indovidually wrapped in Tyvek/plastic blister packages. Plates are plasma etched (TC treated) to support adherent cell lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC3429285 Supplier NEST SCIENTIFIC USA Supplier No. 701001
Add to Cart
Add to Cart
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. This part number comes indovidually wrapped in Tyvek/plastic blister packages. Plates are plasma etched (TC treated) to support adherent cell lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.