Learn More
Alkali Scientific 96-Well Low Adhesion Cell Culture Plates for Suspension Cultures, U-Bottom, Sterile, 1/pk, 100/cs
SDP
Supplier:  Alkali Scientific TL46
Encompass_Preferred
Low adherence Cell Culture Plates­­­­­­­­­­­­­­­­­­­­­­­­­­­ are manufactured in Class 100,000 grade clean room with USP 6 grade raw materials for optimal performance. Suspension cell growth and proliferation in media (not on a surface) can be expected within these low adherence / low adhesion plates. The plates have an overall neutral charge which minimizes cell spreading. A moderately hydrophilic surface for Cell Cultures that effectively inhibit cell adhesion.
Catalog No. 50-238-9446