Learn More
Bellco Glass, Inc. 96 Well Cell Culture Plate, V-bottom, Non-Treated, sterile 1/pk, 100/cs
SDP
Catalog No. 502329331
Encompass_Preferred
96 Well Cell Culture Plate, V-bottom, Non-Treated, sterile 1/pk, 100/cs. cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assure uniform growth. Tissue treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. • Sterilized by E-beam, SAL=10-6.• High clarity, 100% virgin polystyrene.• “TC” refers to vacuum plasma tissue culture treatment.• Clear lot number for batch traceability.• Markings of well coordinates available for 12, 24, 48, 96 and 384 well plates.• Non-Pyrogenic, DNase/Rnase free.
Catalog No. 50-232-9331 Supplier Bellco Glass, Inc. Supplier No. 451400014
96 Well Cell Culture Plate, V-bottom, Non-Treated, sterile 1/pk, 100/cs. cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assure uniform growth. Tissue treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. • Sterilized by E-beam, SAL=10-6.• High clarity, 100% virgin polystyrene.• “TC” refers to vacuum plasma tissue culture treatment.• Clear lot number for batch traceability.• Markings of well coordinates available for 12, 24, 48, 96 and 384 well plates.• Non-Pyrogenic, DNase/Rnase free.