Learn More
Genesee Scientific Corporation Genesee Scientific GenClone 96-Well Cell Culture Plates, Tissue Culture Treated, Gamma Irradiated, 100 Plates/Unit
SDP
Catalog No. NC3245806
Encompass
The Genesee Scientific GenClone 96-Well Cell Culture Plates are constructed from high-quality polystyrene, facilitating reliable cell growth and experimentation. With tissue culture treatment and striation-free, optically clear surfaces, these plates ensure consistent results in microscopy. Certified RNase, DNase, and pyrogen-free, they are manufactured in a Class 100,000 clean room and feature unique lid designs to reduce evaporation and cross-contamination, making them ideal for scientific applications.
Catalog No. NC3245806 Supplier Genesee Scientific Corporation Supplier No. 25109
The Genesee Scientific GenClone 96-Well Cell Culture Plates are constructed from high-quality polystyrene, facilitating reliable cell growth and experimentation. With tissue culture treatment and striation-free, optically clear surfaces, these plates ensure consistent results in microscopy. Certified RNase, DNase, and pyrogen-free, they are manufactured in a Class 100,000 clean room and feature unique lid designs to reduce evaporation and cross-contamination, making them ideal for scientific applications.