Learn More
Diversified Biotech Inc 96-WELL CHAMBER FOR 0.2ML
Catalog No. NC1766924
Encompass
96-well Chamber for 0.2ml PCR plates/tubes.Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures. Dimensions: 127mm x 85mm x 25mm
Catalog No. NC1766924 Supplier Diversified Biotech Inc Supplier No. CHAMA100
96-well Chamber for 0.2ml PCR plates/tubes.Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures. Dimensions: 127mm x 85mm x 25mm