Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
Catalog No. 07000150 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Translucent
White
Yellow
Sterility:
Non-sterile
Sterile
Quantity:
100/Cs.
80/Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Quantity
07-000-150 Translucent Sterile 100/Cs.
07-000-143 Black Non-sterile 100/Cs.
07-000-146 Translucent Non-sterile 100/Cs.
07-000-996 Translucent Sterile 80/Cs.
07-000-144 Blue Non-sterile 100/Cs.
07-000-145 Green Non-sterile 100/Cs.
07-000-147 Red Non-sterile 100/Cs.
07-000-149 Yellow Non-sterile 100/Cs.
07-000-148 White Non-sterile 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 07-000-150 Supplier Greiner Bio-One Supplier No. 650261
Add to Cart
Edge
Add to Cart
For high-throughput screening in cell based test systems.
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 8 x 12
Color Translucent
Product Type Storage Microplate
Material Polypropylene
Volume (Metric) Well 50 to 300 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
Quantity 100/Cs.