Learn More
Greiner Bio-One 96-well μClear™ Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07000614 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-614 Clear
1 options
Catalog No. 07-000-614 Supplier Greiner Bio-One Supplier No. 651162
Add to Cart
Add to Cart
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Clear Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Manufactured from virgin polystyrene
  • Non-cytogenic
TRUSTED_SUSTAINABILITY
Specifications
Array 8 x 12
Packaging Inner bags of 25
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Well Shape Conical Bottom (V-well)
Quantity 375/Cs.
RNase Free Yes