Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates
GREENER_CHOICE

Catalog No. 07000560 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Round
Square, Flat Bottom (F-well)
Volume (Metric) Well:
1.9 μL
3.9 μL
Product Type:
LBR
Standard
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Volume (Metric) Well Product Type
07-000-560 Round 1.9 μL Standard
07-000-566 Round 1.9 μL LBR
07-000-568 Square, Flat Bottom (F-well) 3.9 μL LBR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-560 Supplier Greiner Bio-One Supplier No. 609120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.

  • Non-sterile
  • Polystyrene
  • Good optical properties
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Clear
Product Type Standard
Material Polystyrene
Volume (Metric) Well 1.9 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 40
Quantity 40/Cs.
Height (Metric) 14.4 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.