Learn More
Arcticwhite LLC 96 Round Deep Well, 2 ML, Polypropylene, Round Bottom, Non-sterilized, 96-well, ANSI/SLAS (SBS) Standard Footprint, Medical-grade Polypropylene (PP), Compatible to -86 Deg C, DNase-free, RNase-free, 50 Deep Well Plates per Case
SDP
Catalog No. NC1286831
Encompass
96 Round Deep Well, 2 ML, Polypropylene, Round Bottom, Non-sterilized, 96-well, ANSI/SLAS (SBS) Standard Footprint, Medical-grade Polypropylene (PP), Compatible to -86 Deg C, DNase-free, RNase-free, 50 Deep Well Plates per Case. A 96-well deep well microplate for automated liquid handling, nucleic acid purification, and sample storage. Format - 96-well, ANSI/SLAS (SBS) standard footprint. Well shape - Round. Bottom - Round. Well volume - 2 mL. Material - Medical-grade polypropylene (PP). Temperature range - Compatible to -86 deg C. Certifications - DNase-free, RNase-free. Supplied by Arctic White as 50 deep well plates per case.
Catalog No. NC1286831 Supplier Arcticwhite LLC Supplier No. AWLS219020
96 Round Deep Well, 2 ML, Polypropylene, Round Bottom, Non-sterilized, 96-well, ANSI/SLAS (SBS) Standard Footprint, Medical-grade Polypropylene (PP), Compatible to -86 Deg C, DNase-free, RNase-free, 50 Deep Well Plates per Case. A 96-well deep well microplate for automated liquid handling, nucleic acid purification, and sample storage. Format - 96-well, ANSI/SLAS (SBS) standard footprint. Well shape - Round. Bottom - Round. Well volume - 2 mL. Material - Medical-grade polypropylene (PP). Temperature range - Compatible to -86 deg C. Certifications - DNase-free, RNase-free. Supplied by Arctic White as 50 deep well plates per case.