Learn More
Azzota Corp 96-Well Deep Well Plate 2.3ml
Catalog No. 5000886914
Encompass_Preferred
2 3ml 96-Well Deep Well Plate Square Top shape V-Bottom Sterile PP(Polypropylene) Material 5/pk 50/case for Kingfisher Flex
Catalog No. 50-008-86914 Supplier Azzota Corp Supplier No. AZ96DWP50
2 3ml 96-Well Deep Well Plate Square Top shape V-Bottom Sterile PP(Polypropylene) Material 5/pk 50/case for Kingfisher Flex