Learn More
Labcon North America Labcon PurePlus 96 Well Deep Well Plates, Autoclavable, 2.2 mL Working Volume, 100 Plates per Case
Catalog No. NC1991570
Encompass_Preferred
Labcon PurePlus 96 Well Deep Well Plates are designed for versatile laboratory applications, providing reliable sample handling and storage. These high-quality plates feature 96 autoclavable square wells with a 2.2 mL volume, ensuring contaminant-free conditions for sensitive assays. Made from 100% virgin polypropylene, they offer compatibility with manual and robotic handling systems, while the SBS format and three-sided labeling simplify usage. Each case contains 100 plates, making them ideal for high-throughput environments.
Catalog No. NC1991570 Supplier Labcon North America Supplier No. 39095200009
Add to Cart
Add to Cart
Labcon PurePlus 96 Well Deep Well Plates are designed for versatile laboratory applications, providing reliable sample handling and storage. These high-quality plates feature 96 autoclavable square wells with a 2.2 mL volume, ensuring contaminant-free conditions for sensitive assays. Made from 100% virgin polypropylene, they offer compatibility with manual and robotic handling systems, while the SBS format and three-sided labeling simplify usage. Each case contains 100 plates, making them ideal for high-throughput environments.