Learn More
Alkali Scientific ASI™ 96 Well ELISA Plate, 8-Stripwell, Detachable, High Binding, 50/cs, Black Frame & White Well
SDP
Catalog No. 503027684
Encompass_Preferred
ELISA Plates Stripwells are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. • Conform to SBS/ ANSI standards• plates arrive with low evaporation lids• perfectly suited for multichanneling• maximum well volume of 325ul• Both detachable and undetectable plates are available• Flat and transparent bottom• Excellent well uniformity, CV <5% Material:  optically clear polystyrene (PS), designed for ELISA assays and multichanneling.
Catalog No. 50-302-7684 Supplier Alkali Scientific Supplier No. MP28
Add to Cart
Add to Cart
ELISA Plates Stripwells are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. • Conform to SBS/ ANSI standards• plates arrive with low evaporation lids• perfectly suited for multichanneling• maximum well volume of 325ul• Both detachable and undetectable plates are available• Flat and transparent bottom• Excellent well uniformity, CV <5% Material:  optically clear polystyrene (PS), designed for ELISA assays and multichanneling.