Learn More
Alkali Scientific ASI™ 96 Well ELISA Plate, 8-Stripwell, Detachable, High Binding, 50/cs, White Frame & White Well
SDP
Catalog No. 503026886
Encompass_Preferred
ELISA Plates Stripwells are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. • Conform to SBS/ ANSI standards• plates arrive with low evaporation lids• perfectly suited for multichanneling• maximum well volume of 325ul• Both detachable and undetectable plates are available• Flat and transparent bottom• Excellent well uniformity, CV <5% Material:  optically clear polystyrene (PS), designed for ELISA assays and multichanneling.
Catalog No. 50-302-6886 Supplier Alkali Scientific Supplier No. MP27
Add to Cart
Add to Cart
ELISA Plates Stripwells are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. • Conform to SBS/ ANSI standards• plates arrive with low evaporation lids• perfectly suited for multichanneling• maximum well volume of 325ul• Both detachable and undetectable plates are available• Flat and transparent bottom• Excellent well uniformity, CV <5% Material:  optically clear polystyrene (PS), designed for ELISA assays and multichanneling.