Learn More
Greiner Bio-One 96-well Flat Bottom Chimney Style Non-binding Microplates
GREENER_CHOICE
Catalog No. 07000634 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Color
07-000-634 Black
07-000-636 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-634 Supplier Greiner Bio-One Supplier No. 655900
Add to Cart
Add to Cart
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Flat Bottom Chimney Style Non-binding Microplates are free of detectable DNase, RNase, human DNA.
  • Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
  • Stable under normal assay conditions without degradation or leaching
  • Improves signal-to-noise-ratio
  • Manufactured under DIN ISO 9001 guidel
Service Plan
No
Warranty and Services
No
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
Color Black
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
No. per Pack 10
Quantity 40/Cs.
Well Color Black
Dimensions (L x W x H) 127.8 x 85.5 x 14.4 mm