Learn More
Analytical Sales & Services 96 Well, Flat Bottom Clear PS Microplate, 25-340µL working volume per well
Catalog No. 5000806103
Encompass
96-Well, Flat Bottom Clear Well PS Microplate\n\n• Clear Polystyrene\n• Flat Bottom\n• 25-340µL working volume per well\n• Comes with Lid and Tissue Culture Treatment\n\nQty/Pkg: 100\n\n 
Catalog No. 50-008-06103 Supplier Analytical Sales & Services Supplier No. 96518
96-Well, Flat Bottom Clear Well PS Microplate\n\n• Clear Polystyrene\n• Flat Bottom\n• 25-340µL working volume per well\n• Comes with Lid and Tissue Culture Treatment\n\nQty/Pkg: 100\n\n 