Learn More
Falcon™ 96-Well Flat Bottom TC-Treated Microplate with Lid
Catalog No. 03150863 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
White, Clear Bottom
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-150-863 White, Clear Bottom
03-150-862 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-150-863 Supplier Falcon™ Supplier No. 353389
  • PRODUCT PROMOTION
    $258.00
    $146.34 / Case of 50
    Save $111.66
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls prevent well-to-well cross-talk
  • Optically clear flat bottom permits direct microscopic viewing
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Non-pyrogenic
  • Non-treated polystyrene, standard tissue culture (TC), and Corning™ Primaria™ surface treatments available
  • Convenient, peel-open medical-style packaging for individual product presentation
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
  • Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid Yes
Culture Area 0.34 cm2
Array 8 x 12
Color White, Clear Bottom
Product Type Microplate
Material Virgin Polystyrene
Volume (Metric) Well 0.37 mL
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Well Color Clear