Learn More
Biotix™ 96-Well Format Assay Plates
Catalog No. 12111070 Shop All Biotix Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Round Bottom (U-well)
Sterility:
Non-sterile
Sterile
Total Volume (Metric) Well:
330 μL
350 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Sterility Total Volume (Metric) Well
12-111-070 Conical Bottom (V-well) Non-sterile 330 μL
12-111-071 Conical Bottom (V-well) Sterile 330 μL
12-111-072 Round Bottom (U-well) Non-sterile 350 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-111-070 Supplier Biotix™ Supplier No. 63300102
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Large well diameter with raised Chimney-style wells help reduce cross-contamination.
  • Made from medical grade virgin polypropylene
  • DNase, RNase and Endotoxin (pyrogen) free
  • Alphanumeric grid to help with sample identification and organization
  • Notched bottom corner to be used as a reference for plate orientation
  • Available in V-shaped and U-shaped wells
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
Color Clear
Product Type Microplate
Material Polypropylene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 330 μL
Well Shape Conical Bottom (V-well)
No. per Pack 10
Width (Metric) 85.48 mm
Height (Metric) 14.6 mm