Learn More
Thomas Scientific FrameStar 96 Well Skirted PCR Plate; clear PP wells, clear PC frame, cut corner H1, 50 plates
SDP
Catalog No. NC0510356
Encompass_Preferred
Our FrameStar two-component technology reduces evaporation from PCR plates, improving your results and allowing you to reduce the volume of expensive reagents to save you money. FrameStar plates combine the advantages of thin-walled polypropylene tubes for optimum PCR results, with a rigid polycarbonate frame to increase the thermal stability of the plate during the PCR process. This means that compared to a standard polypropylene plate, the FrameStar plate will not warp and expand during rapid heating and cooling, meaning the integrity of seals will remain consistent and therefore less of your sample will evaporate. This in turn means you can spend less money on expensive reagents, as you are retaining more during the PCR process. Why our FrameStar 2-component design is superior to standard polypropylene plates FrameStar PCR plates maximise thermal stability at high temperatures, which prevents sample loss by minimising thermal expansion during PCR. The two-co
Catalog No. NC0510356 Supplier Thomas Scientific Supplier No. 1149V59
Our FrameStar two-component technology reduces evaporation from PCR plates, improving your results and allowing you to reduce the volume of expensive reagents to save you money. FrameStar plates combine the advantages of thin-walled polypropylene tubes for optimum PCR results, with a rigid polycarbonate frame to increase the thermal stability of the plate during the PCR process. This means that compared to a standard polypropylene plate, the FrameStar plate will not warp and expand during rapid heating and cooling, meaning the integrity of seals will remain consistent and therefore less of your sample will evaporate. This in turn means you can spend less money on expensive reagents, as you are retaining more during the PCR process. Why our FrameStar 2-component design is superior to standard polypropylene plates FrameStar PCR plates maximise thermal stability at high temperatures, which prevents sample loss by minimising thermal expansion during PCR. The two-co