Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 96-well Full Skirt Microplates

Catalog No. 07000619 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-619 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-619 Supplier Greiner Bio-One Supplier No. 652230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with PCR System. Greiner Bio-One 96-Well Full Skirt Microplates are suited for sensitive PCR reactions

  • Manufactured from ultra thin polypropylene for optimal heat transfer
  • Full-skirt feature
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Array 8 x 12
Color Clear
Product Type PCR Microplate
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Quantity 400/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.