Learn More
Biochain Institute Inc 96 Well Human Adult Normal Genomic DNA Plate - Peripheral Blood Leukocyte
SDP
Catalog No. 50179782
Encompass_Preferred
96-Well Genomic DNA plate is specially designed with purified total DNAs (including genomic, mitochondrial and viral DNA) from different donors and different samples including blood, body fluids, tissues, mouse tails, swabs or cultured cells. The genomic DNAs are extracted and purified using BioChain's proprietary techniques that yield high molecular weight material, as verified by agarose gel electrophoresis.
Catalog No. 50-179-782 Supplier Biochain Institute Inc Supplier No. D82341481
96-Well Genomic DNA plate is specially designed with purified total DNAs (including genomic, mitochondrial and viral DNA) from different donors and different samples including blood, body fluids, tissues, mouse tails, swabs or cultured cells. The genomic DNAs are extracted and purified using BioChain's proprietary techniques that yield high molecular weight material, as verified by agarose gel electrophoresis.