Learn More
Biochain Institute Inc 96 Well Breast Tumor Genomic DNA Plate
SDP
Catalog No. 50179783
Encompass_Preferred
96-Well Genomic DNA plate is specially designed with purified total DNAs (including genomic, mitochondrial and viral DNA) from different donors and different samples including blood, body fluids, tissues, mouse tails, swabs or cultured cells. The genomic
Catalog No. 50-179-783 Supplier Biochain Institute Inc Supplier No. D82350861
96-Well Genomic DNA plate is specially designed with purified total DNAs (including genomic, mitochondrial and viral DNA) from different donors and different samples including blood, body fluids, tissues, mouse tails, swabs or cultured cells. The genomic