Learn More
Biochain Institute Inc 96 Well Stomach Tumor Genomic DNA Plate
SDP
Catalog No. 50179786
Encompass_Preferred
BioChain's 96 Well Tumor Genomic DNA Plates can be used as Reference Standard materials for tumor gene expression research. The plate is prepared using the genomic DNAs that are extracted from human tumor tissue samples from as many as 96 individual donor
Catalog No. 50-179-786 Supplier Biochain Institute Inc Supplier No. D82352481
BioChain's 96 Well Tumor Genomic DNA Plates can be used as Reference Standard materials for tumor gene expression research. The plate is prepared using the genomic DNAs that are extracted from human tumor tissue samples from as many as 96 individual donor