Learn More
Chemglass Life Sciences 500uL 96 Well Glass Reaction Plate, Flat Cavity Style
SDP
Catalog No. CG191002
Encompass_Preferred
500uL 96 Well Glass Reaction Plate, Flat Cavity Style. Reaction plate is made of 33-expansion borosilicate glass. Overall dimensions are 85 x 127mm. 500uL. Wells are coded with letters A-H vertically and numbers 1-12 horizontally.
Catalog No. CG191002 Supplier Chemglass Life Sciences Supplier No. CG191002
500uL 96 Well Glass Reaction Plate, Flat Cavity Style. Reaction plate is made of 33-expansion borosilicate glass. Overall dimensions are 85 x 127mm. 500uL. Wells are coded with letters A-H vertically and numbers 1-12 horizontally.